ATE959441110
AstaTech, Inc.
METHYL 4,5-DIBROMO-2-FLUOROBENZOATE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 4,5-dibromo-2-fluorobenzoate
Inchi Code
1S/C8H5Br2FO2/c1-13-8(12)4-2-5(9)6(10)3-7(4)11/h2-3H,1H3
InChI key
GXQSHUARBHYOST-UHFFFAOYSA-N
Purity
95%
Physical Form
White to Yellow Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=111619
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=AB22734
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.