APOH03CD8DE1
Apollo Scientific Ltd
3,6-Difluoro-2-(trifluoromethoxy)benzyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-(bromomethyl)-1,4-difluoro-3-(trifluoromethoxy)benzene
Inchi Code
1S/C8H4BrF5O/c9-3-4-5(10)1-2-6(11)7(4)15-8(12,13)14/h1-2H,3H2
InChI key
MTFPSRAZEPEGFQ-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
95%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid-Clear
Product Link
https://store.apolloscientific.co.uk/product/36-difluoro-2-trifluoromethoxybenzyl-bromide
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/36-difluoro-2-trifluoromethoxybenzyl-bromide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.