APO428498059
Apollo Scientific Ltd
4'-Fluoro-2'-methylacetophenone
About This Item
Iupac Name
1-(4-fluoro-2-methylphenyl)ethanone
Inchi Code
1S/C9H9FO/c1-6-5-8(10)3-4-9(6)7(2)11/h3-5H,1-2H3
InChI key
GCTZPACJEUDZAX-UHFFFAOYSA-N
storage temp.
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/4-fluoro-2-methylacetophenone
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.