AMBH2D6F8700
Ambeed, Inc.
2,3,5-Tribromo-4-methylthiophene
About This Item
Iupac Name
2,3,5-tribromo-4-methylthiophene
Inchi Code
1S/C5H3Br3S/c1-2-3(6)5(8)9-4(2)7/h1H3
InChI key
DTLNXOZXOZCNMX-UHFFFAOYSA-N
1 of 4
This Item | SAB5200988 | SAB5200888 | SAB5200904 |
---|---|---|---|
conjugate unconjugated | conjugate Allophycocyanin conjugate | conjugate unconjugated | conjugate phycoerythrin (R-PE) conjugate |
species reactivity mouse, human, rat | species reactivity mouse, human, rat | species reactivity mouse, human, rat | species reactivity human, mouse, rat |
Gene Information rat ... Grin2a(24409) | Gene Information rat ... Grin2a(24409) | Gene Information rat ... Grin2a(24409) | Gene Information rat ... Grin2a(24409) |
clone S327A-38, monoclonal | clone S327A-38, monoclonal | clone S327-95, monoclonal | clone S327-95, monoclonal |
antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/67869-13-0.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service