Z743884
ART® SoftFit L Reload System
HLT 200L, size 200 μL, hinged rack, non-sterile
Synonym(s):
aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip
About This Item
hinged rack
sterility
non-sterile
feature
HLT 200L
hinged rack
packaging
pkg of 5X960 ea
manufacturer/tradename
Molecular BioProducts 3752-HR
technique(s)
PCR: suitable
size
200 μL
volume
200 μL
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Reload towers are an environmentally-friendly packaging option that reduces plastic waste and storage space.
- Compatible with SoftFit-L empty racks.
- Color-coded inserts allows quick identification of tip volume.
- Free of DNase, RNase, DNA, ATP, and endotoxin.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service