Key Documents
Z743187
Julabo® bath covers
plexiglas cover to fit CORIO CD-1001F
Select a Size
1 EA
€485.00
€485.00
Check Cart for Availability
Select a Size
Change View
1 EA
€485.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
€485.00
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970247
General description
Transparent Plexiglas hood reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the hood prevents contamination of the samples in the bath. The hood can be used in the temperature range of -10°C to +80°C.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)