Key Documents
Z743171
Julabo® bath covers
flat cover to fit Pura 22, with 6 x 115 mm diam openings and set of rings
Select a Size
1 EA
€346.00
€346.00
Estimated to ship on28 April 2025
Select a Size
Change View
1 EA
€346.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
€346.00
Estimated to ship on28 April 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970574
General description
Flat bath cover for PURA 22 water bath, with 6 openings 115 mm diameter and corresponding ring sets. Place beakers or Erlenmeyer flasks directly on the stainless steel base plate.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)