Z740778
Aldrich® Essentials volumetric flask, Class A, meets DIN ISO-1042
capacity 250 mL, stopper joint: 16, tol. 0.150 mL, PE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PE
stopper size
16
Joint
joint ST/NS neckstopper size 16
joint: stopper16
feature
class A meets DIN ISO-1042
packaging
pkg of 12 ea
tol. measuring range
0.150 mL
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Clear borosilicate glass volumetric flask with polyethylene stopper. Class A, meets DIN ISO-1042 specifications. Blue marking spot and capacity marking. White level marking. Flask are certified and either a batch or individual certificate. Calibrated “To Contain”.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service