Skip to Content
Merck
All Photos(1)

Key Documents

Z699195

Trypticase Soy Broth (TSB) Single strength, Bottled Broth

Synonym(s):

broth media, media broth, trypticase soy

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€61.30

€61.30


Check Cart for Availability


Select a Size

Change View
1 EA
€61.30

About This Item

UNSPSC Code:
41111700
NACRES:
NA.74

€61.30


Check Cart for Availability

packaging

bottle of 100 mL

manufacturer/tradename

Maine Mfg 10496707

application(s)

food and beverages

General description

General purpose medium used in qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms. Trypticase Soy Broth – Single Strength complies with the demands of the DIN Norm 10167 for the detection of Escherichia coli serotype 0157:H7 in foods and FDA-BAM for the isolation of enterohemorrhagic Escherichia coli (EHEC). In addition the media conforms to the formula of the US Pharmacopoeia.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service