Key Documents
Z682675
Ace drying flask, pear-shaped, with side indents
capacity 2000 mL, joint: ST/NS 24/40, glass
Synonym(s):
Aldrich® drying flask, pear-shaped, with side indents
Select a Size
1 EA
€193.00
€193.00
Check Cart for Availability
Select a Size
Change View
1 EA
€193.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€193.00
Check Cart for Availability
material
glass
pear-shaped flask
single-neck
Joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Büchi equivalent, 11580
capacity
2000 mL
General description
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)