Key Documents
Z681679
PureSolv Micro vacuum pump/inert gas connection for multiple systems
pkg of 1 ea
Select a Size
1 EA
€648.00
€648.00
Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
Select a Size
Change View
1 EA
€648.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.85
€648.00
Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
material
anodized aluminum frame
stainless steel tubing
packaging
pkg of 1 ea
manufacturer/tradename
Innovative Technologies
General description
Two tees are required per Pure Solv Micro system when ganging vacuum and gas lines.
To connect 2 systems together you need qty 1 of this product
To connect 3 systems together you need qty 2 of this product
To connect 4 systems together you need qty 3 of this product
To connect 5 systems together you need qty 4 of this product
Legal Information
Pure Solv is a trademark of Inert Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)