Skip to Content
Merck
All Photos(1)

Key Documents

Z676861

ART® Softfit L Reload System

maximum volume 20 μL, Tip design ART 20L, Hinged rack, low retention: no

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 960 EA
€902.00

€902.00


Check Cart for Availability


Select a Size

Change View
5 X 960 EA
€902.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
20 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
low retention: no
Hinged rack
Tip design ART 20L
packaging:
pack of 4800 ea (50 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2749-HR

€902.00


Check Cart for Availability

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier
low retention: no
Hinged rack
Tip design ART 20L

packaging

pack of 4800 ea (50 racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2749-HR

technique(s)

PCR: suitable

maximum volume

20 μL

volume

20 μL

Looking for similar products? Visit Product Comparison Guide

General description

Designed to be compatible with LTS, the Molecular Bio Products Softfit L Reload System provides ART barrier pipette tips in color coded inserts by volume size. Choose from five pipette tip volume sizes with the option of low retention technology.

Each individual insert within the Softfit L Reload System is clearly identified with both product number and lot number allowing researchers to enjoy space-saving and environmental benefits of a reload tip system without loosing product traceability.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service