Skip to Content
Merck
All Photos(2)

Key Documents

Z676691

ART® low retention barrier tips

Tip design ART 100E, Reload insert

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 960 EA
€679.00

€679.00


Check Cart for Availability


Select a Size

Change View
5 X 960 EA
€679.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
100 μL
sterility:
sterile
feature:
barrier
Reload insert
Tip design ART 100E
packaging:
pack of 4800 ea (50 inserts of 96 tips)
manufacturer/tradename:
Molecular BioProducts 2065E-05-RI

€679.00


Check Cart for Availability

material

colorless

sterility

sterile

feature

barrier
Reload insert
Tip design ART 100E

packaging

pack of 4800 ea (50 inserts of 96 tips)

manufacturer/tradename

Molecular BioProducts 2065E-05-RI

technique(s)

PCR: suitable

maximum volume

100 μL

volume

100 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service