Skip to Content
Merck
All Photos(1)

Key Documents

Z676012

Dual solution® pipetting reservoir

pack of 25 ea (Pack of 25 reservoirs and 25 lids), non-sterile

Synonym(s):

multichannel pipette reservoir, reagent reservoir

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€91.40

€91.40


Check Cart for Availability


Select a Size

Change View
25 EA
€91.40

About This Item

UNSPSC Code:
41121522
NACRES:
NB.01

€91.40


Check Cart for Availability

material

V-bottom bottom
colorless
polypropylene

sterility

non-sterile

packaging

pack of 25 ea (Pack of 25 reservoirs and 25 lids)

manufacturer/tradename

Heathrow Scientific HS20821A

capacity

250 mL

General description

  • DualSpeed pipetting reservoir comprises polypropylene solution. Basin/reagent reservoir has a standard 50 mL basin on one side and 12 individual 5 mL basins on the other side.
  • Wells are angled both downward and outward, guiding multichannel pipettor tips towards the centered position, helping to avoid contamination and waste.
  • Imprinted indices identify samples: loose fitting lids prevent evaporation and contamination.
  • Innovative shape guides and protects pipette tips
  • Non-sterile, polypropylene

Legal Information

Dual Solution is a registered trademark of Heathrow Scientific
DualSpeed is a trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are the resevoirs autoclavable?

    1 answer
    1. The product is autoclavable.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service