Skip to Content
Merck
All Photos(1)

Key Documents

Z654175

Mettler Toledo SevenGo portable pH meter

SG2-B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

feature

CE compliant

availability

available only in EU
available only in EU

mV measuring range

-1999-1999, accuracy: 1, resolution: 1

pH measuring range

0.00-14.00, accuracy: 0.01, resolution: 0.01

temperature measuring range

-5.0-105 °C, accuracy: 0.5 °C, resolution: 0.1 °C

size

220 mm × 90 mm × 45 mm

weight

325 g

battery power

4 AA

Looking for similar products? Visit Product Comparison Guide

General description

  • Clear display of measured values and settings
  • Continuous indication of electrode and battery status
  • Automatic endpoint and buffer recognition as well as temperature compensation
  • Data memory for 30 GLP data points
  • 3-point calibration with predefined or user-defined buffers
  • Water tight according to IP67
Supplied with a wrist strap, 4 AA batteries, operating instructions, a test certificate and a declaration of conformity

Components

meter including electrode holder

Legal Information

SevenGo is a trademark of Mettler-Toledo, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service