Key Documents
Z615609
Julabo® bath carrier trays for SW22/SW23
to fit 18 Erlenmeyer flasks 100 cc
Select a Size
1 EA
€248.00
€248.00
Estimated to ship on16 April 2025
Select a Size
Change View
1 EA
€248.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.77
€248.00
Estimated to ship on16 April 2025
manufacturer/tradename
Julabo 8970362
Julabo
General description
Erlenmeyer flasks can be positioned in the shaking water bath using the stainless steel tray. A maximum of 18 Erlenmeyer flasks with 100 ml capacity fit on a standard tray.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)