Key Documents
Z607614
Bohlender HT (high temp) laboratory screw joints
GL 14 thread, tubing O.D. 8.0 mm
Select a Size
1 EA
€33.60
€33.60
Check Cart for Availability
Select a Size
Change View
1 EA
€33.60
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€33.60
Check Cart for Availability
manufacturer/tradename
Bohlender
availability
not available in USA and Canada
tubing O.D.
8.0 mm
fitting
GL 14 thread
General description
Pressure resistant up to approx. 10 bar (at room temperature for thread sizes GL 14, GL 18 and GL 25), suitable for vacuum. The circulating medium is exposed only to PTFE; screw cap made of glass-fibre PPS (max.+250 °C). For many years laboratory screw joints have been mainly used for connecting glass tubes with PTFE tubing or with plastic, glass and metal tubes. In addition, these couplings are also suitable for locking probes (even stainless steel), thermometers, laboratory stopcocks, immersion tubes or lead-in cables.
Legal Information
Bohlender is a trademark of Bohlender GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)