Key Documents
Z568694
Sigma-Aldrich
Aldrich® combustion apparatus flask, with stopper and pinch clamp
capacity 1000 mL
Select a Size
1 EA
€439.00
€439.00
Check Cart for Availability
Select a Size
Change View
1 EA
€439.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€439.00
Check Cart for Availability
material
borosilicate glass
conical
single-neck
sterility
non-sterile
feature
closure type Stopper
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
1000 mL
General description
Heavy-wall borosilicate glass conical flask with 35/25 ball joint stopper. Use with a detachable platinum sample carrier suspended from the stopper hook. Pinch clamp holds stopper in place during combustion.
Order platinum carrier and sample wrappers separately.
Based on a design by C.L. Ogg, R.B. Kelly, and J.A. Connelly. See Microchemical Techniques, N.D. Cheronis, Ed. Interscience, 1962, pp 427-433.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)