Z566039
Pure-Pac® II dispense coupler kit
1/2in. tube end
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.01
Pricing and availability is not currently available.
General description
The Sigma-Aldrich Pure-Pac II returnable containers require the Micromatic MacroValve coupler for dispensing chemical. The Dispense Head Coupler Kit includes the MacroValve coupler along with the standard components needed to connect to either 1/2in. or 3/8in. diameter transfer lines. All of the hardware in the kit is supplied in a foam-lined plastic case. See Technical Bulletin AL-236 for assembly instructions.
Legal Information
Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany
Kit Components Also Available Separately
Product No.
Description
SDS
- Z104388PTFE sealing tape, W 1/2 in.SDS
- Adapter for Kilo-Lab® and Pure-Pac® cylinders, ferrule size 1/2 in., NPTM 1/2 in.
- Z560723Micromatic MacroValve coupler for returnable containersSDS
- Swagelok® hex reducing coupling, pkg of 1 ea
- Swagelok® street elbow, pkg of 1 ea
- Z565547Stainless steel hose connector, 1/4in. male NPT, 1/4in. hose I.D., pkg of 1 eaSDS
Choose from one of the most recent versions:
Certificates of Analysis (COA)