Key Documents
Z557447
Julabo® bath spring clamps for Erlenmeyer flasks
fits 100 mL flasks
Select a Size
1 EA
€37.50
€37.50
Estimated to ship on17 April 2025
Select a Size
Change View
1 EA
€37.50
About This Item
UNSPSC Code:
41122413
NACRES:
NB.77
€37.50
Estimated to ship on17 April 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970604
General description
The individual spring clips can be positioned in the shaking water bath using the base tray for spring clips (Z743201-1EA). A maximum of 21 spring clips for 100 ml Erlenmeyer flasks can be mounted on a base tray.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)