Merck
All Photos(6)
Key Documents
Z544701
Molymod® Individual organic model set
Starch or cellulose
Select a Size
1 SET
€25.20
€25.20
Check Cart for Availability
Select a Size
Change View
1 SET
€25.20
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
€25.20
Check Cart for Availability
description
glucose units 3
manufacturer/tradename
Molymod
General description
The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Legal Information
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)