Key Documents
Z544485
Molymod® molecular model set
teacher/researcher set, biochemistry
Select a Size
1 SET
€113.00
€113.00
Check Cart for Availability
Select a Size
Change View
1 SET
€113.00
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
€113.00
Check Cart for Availability
feature
no. of atoms 257
no. of bonds 160
manufacturer/tradename
Molymod MMS-007
General description
100 hydrogen atom-links, 10 hydrogen (ll), 68 carbon (ll, lll, lV), 40 oxygen (l, ll), 34 nitrogen (ll, lll, lV), 2 sulphur (ll), 6 phosphorus (lV), 2 metal (lV, Vl), Links: 150 non-vis colourless & 10 ‘V′-links
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. May be upgraded or customized with additional model parts, for more info, go to sigma-aldrich.com and enter Molymod in our search engine.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)