Skip to Content
Merck
All Photos(1)

Documents

Z529710

Supelco

Aldrich® chromatography sprayer

size 10 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NC.07

grade

for chromatography

technique(s)

thin layer chromatography (TLC): suitable

size

10 mL

Looking for similar products? Visit Product Comparison Guide

General description

Provide a fine, uniform spray that is optimized for the development of TLC plates. Also suited for use in electrophoresis.
  • Adjustable spray pattern using thumb on vent hole
  • Greaseless, screw-threaded joint will not seize; a simple turn of the threaded cap pulls joint apart safely
  • Threaded SafetyBarbs® prevent accidental breakage during tubing installation or removal
  • Uses low-pressure gas or air

Linkage

See the Threaded Ground Glass Tutorial to see how threaded joints work.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evgeni Eltzov et al.
Biosensors & bioelectronics, 87, 572-578 (2016-09-13)
A new colorimetric immunoassay concept, utilizing conventional lateral flow membranes (e.g., conjugation, sample, absorption and nitrocellulose), were placed in a different configuration in a stacking manner, where the liquid sample that may contain the analyte diffuses from the bottom to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service