Key Documents
Z507954
Recovery flask
capacity 10 mL
Select a Size
1 EA
€39.40
€39.40
Check Cart for Availability
Select a Size
Change View
1 EA
€39.40
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€39.40
Check Cart for Availability
material
glass
round bottom flask
single-neck
sterility
non-sterile
feature
closure type Holed cap
packaging
pack of 1
manufacturer/tradename
Ace Glass 9470-43
capacity
10 mL
General description
<ST/>14/10 threaded joint, and modified sides for easy cleaning or removal of material. Supplied with holed cap, PTFE-faced silicone septum, and FETFE® O-ring.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)