Key Documents
Z503371
Sigma-Aldrich
Scienceware® clip clamp
midi
Select a Size
12 EA
€18.40
€18.40
Check Cart for Availability
Select a Size
Change View
12 EA
€18.40
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
€18.40
Check Cart for Availability
feature
for tubing o.d. 3.2 ‑ 11 mm
packaging
pkg of 12 ea
manufacturer/tradename
Bel-Art H18228-0000
General description
Clamps range from the simple on/off position of the mini, to a ratcheting control for the midi and maxi.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What kind of plastic is this product made out of?
    1 answer
    1. This product is an Acetal plastic, also known as polyoxymethylene (POM).
      Helpful?