Z416363
EZSafe® flash-chromatography assembly
With fritted disc (porosity C), capacity 100 mL
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12000000
NACRES:
NC.05
grade
for chromatography
technique(s)
LPLC: suitable
capacity
100 mL
suitability
suitable for chromatography
Looking for similar products? Visit Product Comparison Guide
General description
These columns are improved low-pressure columns (max. pressure 7psig) with unbreakable PTFE fittings and screw-thread connections for increased safety and economy.
The assemblies are supplied with bottom PTFE valve assembly, GL-45 cap, and flow controller. Solvent reservoirs and GL-45 connectors are sold separately.
- Bottom tip breakage is eliminated because the valve and exposed drip-tip is all PTFE
- GL-45 screw-thread connections ensure positive reliable connections
- Eliminates frozen PTFE and glass plugs
- Eliminates grease
- Choose between standard columns or columns with a fritted disc
The assemblies are supplied with bottom PTFE valve assembly, GL-45 cap, and flow controller. Solvent reservoirs and GL-45 connectors are sold separately.
Legal Information
EZ Safe is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service