Key Documents
Z414220
Sigma-Aldrich
Aldrich® septum-inlet adapter, female
Plain, joint: ST/NS 19/22
Select a Size
1 EA
€46.80
€46.80
Check Cart for Availability
Select a Size
Change View
1 EA
€46.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€46.80
Check Cart for Availability
Joint
joint: ST/NS19/22
Related Categories
General description
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)