Key Documents
Z414190
Sigma-Aldrich
Aldrich® pour-out adapter
male joint: ST/NS 29/32
Select a Size
1 EA
€48.80
€48.80
Estimated to ship on30 May 2025
Select a Size
Change View
1 EA
€48.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€48.80
Estimated to ship on30 May 2025
Joint
male joint: ST/NS29/32
General description
For pouring from flasks or funnels without pouring over ground joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)