Key Documents
Z312231
Gerhardt® STL56 warm air glassware dryers
AC/DC input 240 V AC, European 2-pin plug
About This Item
UNSPSC Code:
12352202
Pricing and availability is not currently available.
grade
CE Compliant
feature
Power: 900 W heater
manufacturer/tradename
Gerhardt
availability
not available in USA and Canada
W × D × H
380 mm × 380 mm × 1150 mm
General description
Blows warm air up tubular supports for rapid drying of laboratory glassware. 56 white plastic-coated lateral drying tubes (24, 16 mm diam. 32 tubes, 11 mm diam.) on rotating central column. Electric heater and fan blows filtered hot or cold air up column and out of tube. Maximum temperature in glass vessel on dryer: approx 50 °C.
Heater top has raised lip to act as condensation/drip tray with drain pipe at rear. Front panel has power and heating indicator lamps, power switch, toggle switch for hot or cold air, and timer.
Supplied with Euro 2-pin plug.
Legal Information
Gerhardt is a registered trademark of Gerhardt
Choose from one of the most recent versions:
Certificates of Analysis (COA)