Z303380
Quickfit® double surface (Davies) condenser
L 200 mm, joint: ST/NS 24/29
Synonym(s):
C5/23/SC
Select a Size
€134.00
Estimated to ship on28 May 2025
Select a Size
About This Item
€134.00
Estimated to ship on28 May 2025
Joint
joint: ST/NS24/29
manufacturer/tradename
Quickfit C 5/23/SC
availability
not available in USA and Canada
not available in USA and Canada
L
200 mm
surface area
300 cm2
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
- Highly efficient condenser with large cooling surface making it suitable for distillation, reflux & Soxhlet extraction applications. Suitable for substances with low boiling points.
- Inner cooling surface gives ′baffle′ effect
- Outer cooling surface prevents ′creep′ with low boiling point liquids
- Supplied with detachable plastic screw thread connectors which allow 9mm bore flexible tubing to be fitted easily and safely
- Note: The CX5/25/SC condenser has a plain tube top instead of a socket joint
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service