Key Documents
Z303224
Quickfit® Liebig condenser
L 250 mm, joint: ST/NS 29/32
Synonym(s):
C1/14/25/SC
Select a Size
1 EA
€111.00
€111.00
Estimated to ship on13 April 2025
Select a Size
Change View
1 EA
€111.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€111.00
Estimated to ship on13 April 2025
Joint
joint: ST/NS29/32
manufacturer/tradename
Quickfit C1/14/25/SC
availability
not available in USA and Canada
not available in USA and Canada
L
250 mm
surface area
130 cm2
General description
Designed to give a turbulent flow in the water jacket for efficient cooling at a comparatively slow rate of water flow. These condensers are adequate and effective in many applications.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)