Key Documents
Z302635
Quickfit® round-bottom flasks
capacity 25 mL, joint: ST/NS 19/26
Synonym(s):
FR25/2S
Select a Size
1 EA
€23.10
5 EA
€85.80
€23.10
Estimated to ship on28 May 2025
Select a Size
Change View
1 EA
€23.10
5 EA
€85.80
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€23.10
Estimated to ship on28 May 2025
material
borosilicate glass
round bottom flask
short
single-neck
Joint
joint: ST/NS19/26
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
case of 10
manufacturer/tradename
Quickfit FR25/2S
availability
not available in USA and Canada
capacity
25 mL
General description
Short neck with ground-glass joint.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)