Key Documents
Z301310
Quickfit® plain bends
joint: ST/NS 19/26
Synonym(s):
RA9/22
Select a Size
1 EA
€44.50
€44.50
Estimated to ship on30 May 2025
Select a Size
Change View
1 EA
€44.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€44.50
Estimated to ship on30 May 2025
Joint
joint: ST/NS19/26
manufacturer/tradename
Quickfit RA 9/22
availability
not available in USA and Canada
not available in USA and Canada
General description
Socket to cone. Generally used as distilling adapters.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)