Key Documents
Z301175
Quickfit® expansion adapter
top female joint: ST/NS 34/35, bottom male joint: ST/NS 29/32
Synonym(s):
XA54
Select a Size
1 EA
€55.80
€55.80
Estimated to ship on16 April 2025
Select a Size
Change View
1 EA
€55.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€55.80
Estimated to ship on16 April 2025
Joint
female joint: ST/NS top34/35
male joint: ST/NS bottom29/32
manufacturer/tradename
Quickfit XA 54
availability
not available in USA and Canada
not available in USA and Canada
General description
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)