Key Documents
Z300926
Quickfit® Reduction Adapters
top female joint: ST/NS 19/26, bottom male joint: ST/NS 29/32
Synonym(s):
DA24
Select a Size
1 EA
€27.60
€27.60
Check Cart for Availability
Select a Size
Change View
1 EA
€27.60
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€27.60
Check Cart for Availability
Joint
female joint: ST/NS top19/26
male joint: ST/NS bottom29/32
manufacturer/tradename
Quickfit DA 24
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)