Key Documents
Z266051
Supelco
PTFE multi-plate racks
For use with 20 cm × 20 cm plates
Select a Size
1 EA
€309.00
€309.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
€309.00
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
€309.00
Check Cart for Availability
Request a Bulk Order
technique(s)
thin layer chromatography (TLC): suitable
General description
For use with Latch-Lid developing chamber. Can accommodate up to six TLC plates.
Legal Information
Latch-Lid is a trademark of General Glassblowing, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Christiane Mühle et al.
Journal of chromatography. A, 1481, 137-144 (2016-12-26)
As part of the sphingomyelin pathway, sphingomyelinases and ceramidases have attracted much attention in basic as well as clinical research. However, current assays still often rely on a radioactive substrate, extensive manual purification steps, and hazardous solvents for chromatographic analysis.