Key Documents
Z261491
Luer-to-Tubing Connector
bulkhead panel mounted Micro-Mate® female Luer to hose end for 1/16 in. to 3/32 in. I.D. tubing, nickel plated
Synonym(s):
environmental, veterinary
Select a Size
1 EA
€54.70
€54.70
Check Cart for Availability
Select a Size
Change View
1 EA
€54.70
About This Item
UNSPSC Code:
40183110
NACRES:
NB.03
€54.70
Check Cart for Availability
material
nickel plated
manufacturer/tradename
Popper 6421
General description
Hardware specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. 316 SS fittings are noncontaminating, easy to clean and resistant to corrosion.
Chrome-plated brass connectors offer good performance at an economical price. Water tested at 45 psi.
Legal Information
Micro-Mate is a registered trademark of Cadence Science
Choose from one of the most recent versions:
Certificates of Analysis (COA)