Z260126
FORTUNA® Optifix® Solvent bottle-top dispenser
Solvents model, volume 2-10 mL
Synonym(s):
Bottle-Top Dispenser, Liquid Dispensing System, Solvent Dispenser
Select a Size
€400.00
Select a Size
About This Item
€400.00
Recommended Products
feature
subdivision 0.2 mL
manufacturer/tradename
Fortuna Graf 101.081-38
volume measuring range
2-10 mL
volume
2-10 mL
fitting
GL 40
GL 45
Looking for similar products? Visit Product Comparison Guide
General description
Features:
- Dispensing piston made of glass
- Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
- Fully positional FIX adapter made of white PP for dispensers up to 10 mL or solid white PTFE for dispensers from 30 - 500 mL size
- Valve block made of PTFE, no metal springs in the valve - no corrosion
- Ergonomic finger protection cap
- Fast and easy volume setting with additional fine adjustment
- Optifix Solvent dispenser available up to 500 mL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service