Key Documents
Z248096
Sigma-Aldrich
Nalgene® hand-operated vacuum pump
PVC body
Synonym(s):
hand vacuum pump, liquid dispensing device
Select a Size
1 EA
€322.00
€322.00
Check Cart for Availability
Select a Size
Change View
1 EA
€322.00
About This Item
UNSPSC Code:
41105100
NACRES:
NB.79
€322.00
Check Cart for Availability
material
PVC body
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 6132-0010
General description
Pump with vacuum gauge attains and holds vacuum of 125 mm Hg (absolute). Vacuum trigger release requires only one hand to operate. Adjustable vacuum release rate controlled by lever on pump. Full blank port pressure obtained in two strokes.
Removable cover on exhaust port and trigger releases vacuum without disconnecting pump from line. Gauge monitors vacuum in cm and in. Hg.
Includes: 2 ft of 1/4 in. I.D. Nalgene 180 tubing.
Use for siphoning, filtration, liquid transfer, bleeding fluid lines, checking for equipment leaks, and adjusting vacuum-operated devices. When transferring liquids, use air trap to prevent liquid from entering pump.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)