Key Documents
Z239089
Pipette fillers
red silicone bulb type
Select a Size
1 EA
€45.60
€45.60
Check Cart for Availability
Select a Size
Change View
1 EA
€45.60
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€45.60
Check Cart for Availability
material
red silicone bulb
manufacturer/tradename
SILRED-001
General description
Valved, choice of natural or silicone rubber bulb. Allows safe manual control of filling and discharging of liquids by finger pressure.
Choose from one of the most recent versions:
Certificates of Analysis (COA)