Z231541
Duroplan petri dishes
O.D. × H 120 mm × 20 mm
Synonym(s):
217555106
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
sterility
non-sterile
feature
lid
packaging
pack of 1 ea
case of 10 ea
manufacturer/tradename
DURAN® 217555106
availability
not available in USA and Canada
O.D. × H
120 mm × 20 mm
dish capacity
~220 mL
dish diam.
120 mm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Supplied in pairs as dish and cover. These petri dishes are manufactured to give even distribution of culture media and distortion-free transparency. Meet DIN 12339 standard.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service