Z223328
Multi-purpose distillation head
Overall H 430 mm, joint: ST/NS 24/40
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS24/40
O.D.
48 mm
Overall H
430 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
With vacuum take-off and anti-splash design. Useful as a concentrator, condenser, reflux head, or addition funnel. 4 mm PTFE stopcock.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service