Key Documents
Z219967
Sigma-Aldrich
Aldrich® hollow pennyhead stopper
joint: ST/NS 45/50
Synonym(s):
glass stopper, glass stoppers, stoppers
Select a Size
1 EA
€89.40
€89.40
Estimated to ship on13 April 2025
Select a Size
Change View
1 EA
€89.40
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
€89.40
Estimated to ship on13 April 2025
material
ground glass
Joint
joint: ST/NS45/50
description
closed at bottom
General description
Conforms to standard-taper specifications as outlined in ASTM E 675.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)