Z214825
Aldrich® modified Davies condenser
jacket L 400 mm, joint: ST/NS 24/40
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS24/40
jacket L
400 mm
Related Categories
General description
Additional internal cooling surface increases efficiency with negligible hold-up when used at a standard 75° angle.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC