Key Documents
Z191388
Sigma-Aldrich
Adjustable Jacobs® chuck
Fits 3/8in. diam. motor shaft
Select a Size
1 EA
€85.30
€85.30
Check Cart for Availability
Select a Size
Change View
1 EA
€85.30
About This Item
UNSPSC Code:
41103801
NACRES:
NB.72
€85.30
Check Cart for Availability
feature
Fits 3/8in. diam. motor shaft
General description
Accepts 1/16 to 3/8in. diam. stirrer rods.
Legal Information
Jacobs is a registered trademark of Jacobs
Choose from one of the most recent versions:
Certificates of Analysis (COA)