Key Documents
Z177849
Scienceware® Ellipso-Spoon® sampler
L 15 cm
Synonym(s):
curved laboratory spatula, laboratory spoon
Select a Size
1 EA
€38.50
€38.50
Check Cart for Availability
Select a Size
Change View
1 EA
€38.50
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
€38.50
Check Cart for Availability
manufacturer/tradename
Bel-Art H368060015
L
15 cm
General description
Special curved shape permits contact with all corners of vessels, bottles, or containers. No corner out of reach and no loss of materials. Handles crystals, powders, granulated and solid substances, even thick liquids or delicate substances. SS, will not bend. Has both spoon end and spatula end.
Legal Information
Ellipso-Spoon is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)