Z171727
Vacuum-jacketed filter funnel
200 mL, 50 mm frit, 25 to 50 μm porosity
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Aldrich
capacity
200 mL
200 mL
pore size
25-50 μm
Related Categories
General description
200 mL, 50 mm frit, 25 to 50 μm porosity. Useful for low-temperature filtrations. 45/50F top joint, 24/40M bottom joint. Includes 45/50M to 24/40F reducing adapter for top joint.