Skip to Content
Merck
All Photos(1)

Key Documents

Z167258

Sigma-Aldrich

Suba-Seal® septa

Suba-Seal® septa
2 of 2 reviewers received a sample product or took part in a promotion

white rubber, Suba-Seal® 13, fits neck I.D., 9.5 mm, for use with 10/30 joints

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€90.70

€90.70


Check Cart for Availability


Select a Size

Change View
100 EA
€90.70

About This Item

UNSPSC Code:
41121800
NACRES:
NB.35

€90.70


Check Cart for Availability

material

white rubber

feature

fits neck I.D. 9.5 mm
for use with 10/30 joints

manufacturer/tradename

Suba-Seal® 13

Looking for similar products? Visit Product Comparison Guide

General description

Well-known as the best quality rubber turn-over closure with serrated stopper, Suba-Seal brand is available exclusively from Sigma-Aldrich. Ideal for air and moisture-sensitive chemistry procedures. After piercing with non-coring needle, rubber closes puncture, providing air and moisture tight seal to protect contents of vessel or container from atmosphere.

This closure has turnover flange molded to grip outside of container neck promoting a double seal in conjunction with annular serrations on hollow plug. Annular serrations depress themselves against the inside wall of the bottle neck, making each serration a suctions sealing point.

Storage Note for Natural Rubber Septa: Store septa in a closed bag out of sunlight to retard blooming, which is the formation of a whitish film on the surface of the rubber. Blooming is a natural process and does not affect performance. Remove visible bloom by washing or wiping off before use.

Note: For extra security when using large bore transfer needles, fasten septum to bottle or flask opening with a wrap-it tie. See ties and accessory tool listings.

Legal Information

Suba-Seal is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

2 of 2 reviewers received a sample product or took part in a promotion

Active Filters

  1. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service