Key Documents
Z166162
Sigma-Aldrich
Aldrich® antisplash splash-guard adapter with return hole
female joint: ST/NS 24/40, male joint: ST/NS 14/20, capacity 250 mL
Select a Size
1 EA
€157.00
€157.00
Estimated to ship on30 May 2025
Select a Size
Change View
1 EA
€157.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
€157.00
Estimated to ship on30 May 2025
Joint
female joint: ST/NS24/40
male joint: ST/NS14/20
capacity
250 mL
General description
These traps greatly reduce carry-over from foaming or bumping. Anything that may climb can return back down through the return hole at the bottom of this adapter.
Application
Aldrich antisplash splash-guard adapter with return hole may be used between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into the condenser in case of "bumping". A drain hole at the base of the inner tube allows solvent to drain back into the evaporation flask.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)