Key Documents
Z153079
Sigma-Aldrich
All-purpose utility shears
autoclavable, pkg of 1 ea
Synonym(s):
multi purpose shears
Select a Size
1 EA
€11.20
€11.20
Check Cart for Availability
Select a Size
Change View
1 EA
€11.20
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
€11.20
Check Cart for Availability
feature
autoclavable
packaging
pkg of 1 ea
General description
SS blades (one serrated) with plastic handles. Autoclavable to 120 °C. Cuts a variety of materials from wire to bone.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2